Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W395218

Sigma-Aldrich

trans-2,cis-6-Nonadienyl acetate

mixture of isomers, ≥97%

Synonym(s):

Nonadienyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH=CHCH2CH2CH=CHCH2OCOCH3
CAS Number:
Molecular Weight:
182.26
FEMA Number:
3952
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

grade

Kosher

assay

≥97%

refractive index

n20/D 1.4530 (lit.)

bp

75 °C/15 mmHg (lit.)

density

0.897 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

apple; green; green; oily

SMILES string

[H]\C(CC)=C(/[H])CC\C([H])=C(/[H])COC(C)=O

InChI

1S/C11H18O2/c1-3-4-5-6-7-8-9-10-13-11(2)12/h4-5,8-9H,3,6-7,10H2,1-2H3/b5-4-,9-8+

InChI key

UHONGPVFPQQOSO-FTGFODROSA-N

Biochem/physiol Actions

Odor at 1%
Taste at 1-15 ppm

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

199.9 °F - closed cup

flash_point_c

93.3 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 25, 72-72 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service