Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W389323

Sigma-Aldrich

2-Pentyl butyrate

>98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H7CO2CH(CH3)C3H7
CAS Number:
Molecular Weight:
158.24
FEMA Number:
3893
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.658

biological source

synthetic

grade

Kosher

assay

>98%

mol wt

158.24 g/mol

color

colorless

refractive index

n20/D 1.4080

bp

58-59 °F/~9 mmHg

application(s)

flavors and fragrances

organoleptic

fatty; sweet

SMILES string

CCCC(C)OC(=O)CCC

InChI

1S/C9H18O2/c1-4-6-8(3)11-9(10)7-5-2/h8H,4-7H2,1-3H3

InChI key

DJOCFLQKCMWABC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

73.4 °F

flash_point_c

23 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service