Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W387001

Sigma-Aldrich

3-Methyl-2-oxopentanoic acid sodium salt

≥98%

Synonym(s):

2-Keto-3-methylvaleric acid sodium salt, 3-Methyl-2-oxopentanoic acid sodium salt, Ketoisoleucine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)COCO2Na
CAS Number:
Molecular Weight:
152.12
FEMA Number:
3870
EC Number:
MDL number:
UNSPSC Code:
12164502

biological source

synthetic

assay

≥98%

mp

204-206 °C (lit.)

SMILES string

[Na+].CCC(C)C(=O)C([O-])=O

InChI

1S/C6H10O3.Na/c1-3-4(2)5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1

InChI key

SMDJDLCNOXJGKC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Coitinho et al.
Brain research, 894(1), 68-73 (2001-03-14)
Neurological dysfunction is common in patients with maple syrup urine disease (MSUD). However, the mechanisms underlying the pathophysiology of this disorder are poorly known. In the present study we investigated the effect of intrastriatal administration of the alpha-keto acids accumulating
Hsueh-Meei Huang et al.
Biochimica et biophysica acta, 1637(1), 119-126 (2003-01-16)
Mitochondrial dysfunction occurs in many neurodegenerative diseases. The alpha-ketoglutarate dehydrogenase complex (KGDHC) catalyzes a key and arguably rate-limiting step of the tricarboxylic acid cycle (TCA). A reduction in the activity of the KGDHC occurs in brains and cells of patients
Carmen Martínez-Cuesta et al.
Biotechnology letters, 25(8), 599-602 (2003-07-29)
The bacteriocin, lacticin 3147, increased isoleucine transamination by Lactococcus lactis IFPL359 in a cheese model system. The formation of alpha-keto-beta-methyl-n-valeric acid and 2-hydroxy-3-methyl-valeric acid increased by three times in cheese slurries at 12 degrees C and cheese aroma intensity increased
Hsueh-Meei Huang et al.
Annals of the New York Academy of Sciences, 1042, 272-278 (2005-06-21)
The alpha-ketoglutarate dehydrogenase complex (KGDHC) is a mitochondrial enzyme in the TCA cycle. Inhibition of KGDHC activity by alpha-keto-beta-methyl-n-valeric acid (KMV) is associated with neuron death. However, the effect of KMV in microglia is unclear. Therefore, we investigated the effect
Cláudia Funchal et al.
Cellular and molecular neurobiology, 26(1), 67-79 (2006-04-25)
Accumulation of the branched-chain alpha-keto acids (BCKA), alpha-ketoisocaproic acid (KIC), alpha-keto-beta-methylvaleric acid (KMV), and alpha-ketoisovaleric acid (KIV) and their respective branched-chain alpha-amino acids (BCAA) in tissues and biological fluids is the biochemical hallmark of patients affected by the neurometabolic disorder

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service