Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W380318

Sigma-Aldrich

Whiskey lactone

≥98%

Synonym(s):

(±)-5-Butyl-4-methyldihydro-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
FEMA Number:
3803
EC Number:
Council of Europe no.:
10535
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.053
Pricing and availability is not currently available.

biological source

synthetic

description

mixture of cis and trans

assay

≥98%

refractive index

n20/D 1.4454 (lit.)

bp

93-94 °C/5 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

coconut; green

SMILES string

CCCCC1OC(=O)CC1C

InChI

1S/C9H16O2/c1-3-4-5-8-7(2)6-9(10)11-8/h7-8H,3-6H2,1-2H3

InChI key

WNVCMFHPRIBNCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service