Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W371424

Sigma-Aldrich

Methyl sorbate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCO2CH3
CAS Number:
Molecular Weight:
126.15
FEMA Number:
3714
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.3

assay

≥97%

refractive index

n20/D 1.503 (lit.)

bp

180 °C (lit.)

density

0.968 g/mL at 25 °C (lit.)

SMILES string

[H]\C(C)=C([H])/C([H])=C(\[H])C(=O)OC

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

158.0 °F - closed cup

flash_point_c

70 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service