Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W362001

Sigma-Aldrich

4,5-Dimethyl-2-ethyl-3-thiazoline, mixture of isomers

97%

Synonym(s):

2-Ethyl-2,5-dihydro-4,5-dimethylthiazole, 2-Ethyl-4,5-dimethyl-3-thiazoline, 2-Ethyl-4,5-dimethylthiazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NS
CAS Number:
Molecular Weight:
143.25
FEMA Number:
3620
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.030
organoleptic:
chocolate; coffee; green; meaty; musty; nutty; peach; sulfurous
grade:
Kosher
biological source:
synthetic
Pricing and availability is not currently available.

biological source

synthetic

grade

Kosher

assay

97%

refractive index

n20/D 1.496

density

0.971 g/mL at 25 °C

application(s)

flavors and fragrances

organoleptic

chocolate; coffee; green; meaty; musty; nutty; peach; sulfurous

SMILES string

CCC1SC(C)C(C)=N1

InChI

1S/C7H13NS/c1-4-7-8-5(2)6(3)9-7/h6-7H,4H2,1-3H3

InChI key

CSACPVARWHDAET-UHFFFAOYSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

150.1 °F - closed cup

flash_point_c

65.6 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service