Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W337218

Sigma-Aldrich

2-(1-Methylpropyl)thiazole

≥98%

Synonym(s):

2-sec-Butylthiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NS
CAS Number:
Molecular Weight:
141.23
FEMA Number:
3372
EC Number:
Council of Europe no.:
11598
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.022

biological source

synthetic

grade

Halal
Kosher

assay

≥98%

refractive index

n20/D 1.499 (lit.)

density

1.001 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

chocolate; green; musty; minty; vegetable; nutty; pepper; sulfurous

SMILES string

CCC(C)c1nccs1

InChI

1S/C7H11NS/c1-3-6(2)7-8-4-5-9-7/h4-6H,3H2,1-2H3

InChI key

MHJSWOZJMPIGJQ-UHFFFAOYSA-N

Biochem/physiol Actions

Taste at 1 ppm

Other Notes

Natural occurrence: Tomato.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

138.2 °F - closed cup

flash_point_c

59 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 58-58 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service