All Photos(1)
About This Item
Linear Formula:
H2C=C(CH3)CH2CH2COCH3
CAS Number:
Molecular Weight:
112.17
FEMA Number:
3365
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.100
biological source
synthetic
assay
99%
refractive index
n20/D 1.431 (lit.)
bp
148-149 °C (lit.)
density
0.865 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
organoleptic
vegetable
SMILES string
CC(=C)CCC(C)=O
InChI
1S/C7H12O/c1-6(2)4-5-7(3)8/h1,4-5H2,2-3H3
InChI key
VBCIOOKAKHGVMI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service