Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W332909

Sigma-Aldrich

S-Allyl thiopropionate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COSCH2CH=CH2
CAS Number:
Molecular Weight:
130.21
FEMA Number:
3329
Council of Europe no.:
11436
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.101
Pricing and availability is not currently available.

assay

≥95%

refractive index

n20/D 1.482 (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)SCC=C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

118.4 °F - closed cup

flash_point_c

48 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service