Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W323101

Sigma-Aldrich

2-(Methylthio)pyrazine

≥96.5%

Synonym(s):

Methyl pyrazinyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2S
CAS Number:
Molecular Weight:
126.18
FEMA Number:
3231
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.034
Pricing and availability is not currently available.

assay

≥96.5%

SMILES string

CSC1=CN=CC=N1

InChI

1S/C5H6N2S/c1-8-5-4-6-2-3-7-5/h2-4H,1H3

InChI key

KBPBOWBQRUXMFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service