W315702
Ethyl (p-tolyloxy)acetate
≥98%
Synonym(s):
Ethyl p-cresoxyacetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C6H4OCH2CO2C2H5
CAS Number:
Molecular Weight:
194.23
FEMA Number:
3157
EC Number:
Council of Europe no.:
2243
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.797
assay
≥98%
refractive index
n20/D 1.502 (lit.)
bp
120-121 °C/1 mmHg (lit.)
density
1.078 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)COc1ccc(C)cc1
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service