Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W313801

Sigma-Aldrich

3,4-Dimethoxystyrene

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH=CH2
CAS Number:
Molecular Weight:
164.20
FEMA Number:
3138
EC Number:
MDL number:
Flavis number:
4.040
Pricing and availability is not currently available.

biological source

synthetic

grade

technical grade

refractive index

n20/D 1.571 (lit.)

bp

120-125 °C/10 mmHg (lit.)

density

1.109 g/mL at 25 °C (lit.)

organoleptic

green

SMILES string

COc1ccc(C=C)cc1OC

InChI

1S/C10H12O2/c1-4-8-5-6-9(11-2)10(7-8)12-3/h4-7H,1H2,2-3H3

InChI key

NJXYTXADXSRFTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service