Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W305308

Sigma-Aldrich

Terpinyl propionate

≥95%, FCC

Synonym(s):

p-Menth-1-en-8-yl-propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O2
CAS Number:
Molecular Weight:
210.31
FEMA Number:
3053
EC Number:
Council of Europe no.:
423
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.142

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FCC
FDA 21 CFR 172.515

assay

≥95%

refractive index

n20/D 1.464 (lit.)

bp

240 °C (lit.)

density

0.949 g/mL at 25 °C (lit.)

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CCC(=O)OC(C)(C)C1CCC(C)=CC1

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

160.0 °F - closed cup

flash_point_c

71.11 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service