About This Item
grade
Kosher
assay
≥95%
optical activity
[α]25/D −6°, neat
refractive index
n20/D 1.456 (lit.)
bp
104-105 °C/10 mmHg (lit.)
density
0.925 g/mL at 25 °C (lit.)
SMILES string
CC1CCC(C(C1)OC(C)=O)C(C)=C
Looking for similar products? Visit Product Comparison Guide
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
186.8 °F - closed cup
flash_point_c
86 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service