Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W295507

Sigma-Aldrich

Propyl phenylacetate

≥99%

Synonym(s):

n-Propyl-α-toluate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
178.23
FEMA Number:
2955
EC Number:
Council of Europe no.:
229
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.702

biological source

synthetic

assay

≥99%

refractive index

n20/D 1.492 (lit.)

bp

240 °C/753 mmHg (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

CCCOC(=O)Cc1ccccc1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service