About This Item
Recommended Products
biological source
synthetic
assay
98%
refractive index
n20/D 1.368 (lit.)
bp
68 °C (lit.)
density
0.884 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
SMILES string
[H]C(=O)OC(C)C
InChI
1S/C4H8O2/c1-4(2)6-3-5/h3-4H,1-2H3
InChI key
RMOUBSOVHSONPZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system, Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
7.0 °F - closed cup
flash_point_c
-13.9 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service