Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W287901

Sigma-Aldrich

4-Phenyl-2-butanol

97%

Synonym(s):

α−Methyl-benzenepropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
150.22
FEMA Number:
2879
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.036

biological source

synthetic

assay

97%

form

liquid

refractive index

n20/D 1.5140 (lit.)

bp

132 °C/14 mmHg (lit.)

density

0.970 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

peony

SMILES string

CC(O)CCc1ccccc1

InChI

1S/C10H14O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-6,9,11H,7-8H2,1H3

InChI key

GDWRKZLROIFUML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service