W287202
Phenoxyacetic acid
≥98%
Synonym(s):
Glycolic acid phenyl ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5OCH2CO2H
CAS Number:
Molecular Weight:
152.15
FEMA Number:
2872
Beilstein/REAXYS Number:
907949
EC Number:
Council of Europe no.:
2005
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.049
grade
Kosher
assay
≥98%
mp
98-100 °C (lit.)
SMILES string
OC(=O)COc1ccccc1
InChI
1S/C8H8O3/c9-8(10)6-11-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)
InChI key
LCPDWSOZIOUXRV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Maria G Margariti et al.
Therapeutic drug monitoring, 29(2), 150-163 (2007-04-10)
Extensive use of synthetic pesticides for agricultural and nonagricultural purposes began in the past 50 years. As a result of their wide and extensive application, exposure to hazardous pesticides is a concern to the general population and occupationally exposed persons.
Katarzyna Ignatowicz
Journal of hazardous materials, 169(1-3), 953-957 (2009-05-19)
This paper presents research on phenoxyacid pesticides removal using sorption methods on activated carbons. It was noted, that physico-chemical properties of adsorbent and adsorbate as well as parameters of the process have influence on adsorption of pesticides, derivatives of phenoxyacetic
Guan-Sheng Jiao et al.
Bioorganic & medicinal chemistry letters, 22(6), 2242-2246 (2012-02-22)
Four core structures capable of providing sub-nanomolar inhibitors of anthrax lethal factor (LF) were evaluated by comparing the potential for toxicity, physicochemical properties, in vitro ADME profiles, and relative efficacy in a rat lethal toxin (LT) model of LF intoxication.
Giuseppe Fracchiolla et al.
ChemMedChem, 2(5), 641-654 (2007-03-16)
Peroxisome proliferator-activated receptors (PPARs) are ligand-activated transcription factors that govern lipid and glucose homeostasis, and play a central role in cardiovascular disease, obesity, and diabetes. Thus, there is significant interest in developing new and specific agonists for these receptors. Herein
Huiting Zhang et al.
Bioorganic & medicinal chemistry, 15(18), 6089-6095 (2007-07-07)
Molecular imprinted polymers (MIPs) binding with phenoxyacetic acid (PA) as a dummy template molecule were synthesized via thermal initiation in aqueous medium. The retention behaviors of benzoic acid (BA), PA, 2-methyl-4-chlorophenoxyacetic acid (MCPA), 4-chlorophenoxyacetic acid (4-CPA), and 2,4-dichlorophenoxyacetic acid (2,4-D)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service