About This Item
biological source
synthetic
grade
Kosher
assay
≥92%
contains
(may contain up to 0.10% alpha-tocopherol, synthetic as stabilzer)
refractive index
n20/D 1.458 (lit.)
bp
252 °C (lit.)
density
0.89 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
organoleptic
ethereal; sage; sweet
SMILES string
CC(C)CC(=O)OC\C=C(\C)CC\C=C(/C)C
InChI
1S/C15H26O2/c1-12(2)7-6-8-14(5)9-10-17-15(16)11-13(3)4/h7,9,13H,6,8,10-11H2,1-5H3/b14-9-
InChI key
SOUKTGNMIRUIQN-ZROIWOOFSA-N
Other Notes
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service