Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W266558

Sigma-Aldrich

L-Menthol crystals

Synonym(s):

(1R,2S,5R)-(−)-Menthol, (−)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
FEMA Number:
2665
Beilstein/REAXYS Number:
1902293
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

grade

Halal
Kosher

vapor pressure

0.8 mmHg ( 20 °C)

bp

212 °C (lit.)

mp

41-45 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

Inchi Key

NOOLISFMXDJSKH-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural Occurrence: Mentha species.

Biochem/physiol Actions

Taste at 25 ppm

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

201.2 °F - closed cup

flash_point_c

94 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hjalte H Andersen et al.
Pain, 156(5), 880-889 (2015-02-27)
Topical high-concentration L-menthol is the only established human experimental pain model to study mechanisms underlying cold hyperalgesia. We aimed at investigating the combinatorial effect of cold stimuli and topical L-menthol on cold pain and secondary mechanical hyperalgesia. Analogue to the
Mosciano, G.
Perfumer & Flavorist, 30, 55-55 (2005)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 77-77 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service