Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W254201

Sigma-Aldrich

1,1-Dimethoxyheptane

98%

Synonym(s):

Heptanal dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OCH3)2
CAS Number:
Molecular Weight:
160.25
FEMA Number:
2542
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.028

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

assay

98%

refractive index

n20/D 1.4105 (lit.)

bp

183 °C/760 mmHg (lit.)

density

0.832 g/mL at 25 °C (lit.)

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CCCCCCC(OC)OC

InChI

1S/C9H20O2/c1-4-5-6-7-8-9(10-2)11-3/h9H,4-8H2,1-3H3

InChI key

BBMCNYFBAIUERL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

136.0 °F - closed cup

flash_point_c

57.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service