Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W242306

Sigma-Aldrich

Ethyl benzoylacetate

≥90%

Synonym(s):

Benzoylacetic acid ethyl ester, Ethyl 3-oxo-3-phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2COOC2H5
CAS Number:
Molecular Weight:
192.21
FEMA Number:
2423
Beilstein/REAXYS Number:
389944
EC Number:
Council of Europe no.:
627
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.476

vapor density

6.6 (vs air)

assay

≥90%

availability

available only in EU

refractive index

n20/D 1.52 (lit.)

bp

265-270 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccccc1

InChI

1S/C11H12O3/c1-2-14-11(13)8-10(12)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

GKKZMYDNDDMXSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

284.0 °F - closed cup

flash_point_c

140 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service