Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W217700

Sigma-Aldrich

Isobutyl acetoacetate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOCH2CH(CH3)3
CAS Number:
Molecular Weight:
158.19
FEMA Number:
2177
Beilstein/REAXYS Number:
1761405
EC Number:
Council of Europe no.:
242
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.404

assay

≥98%

refractive index

n20/D 1.424 (lit.)

bp

100 °C/22 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(=O)CC(C)=O

InChI

1S/C8H14O3/c1-6(2)5-11-8(10)4-7(3)9/h6H,4-5H2,1-3H3

InChI key

ZYXNLVMBIHVDRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Sweet, wine-like, fruity

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

172.4 °F - closed cup

flash_point_c

78 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service