Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W213748

Sigma-Aldrich

Benzyl alcohol

≥99%, JSFA

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
FEMA Number:
2137
Beilstein/REAXYS Number:
878307
EC Number:
Council of Europe no.:
58
MDL number:
Flavis number:
2.010
Pricing and availability is not currently available.

biological source

synthetic

agency

JSFA

reg. compliance

FDA 21 CFR 172.515
FDA 21 CFR 175.105
FDA 21 CFR 175.300
FDA 21 CFR 177.1210
FDA 21 CFR 73.1001

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

assay

≥99%

autoignition temp.

817 °F

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)
205 °C

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

rose

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

213.8 °F - DIN 51758

flash_point_c

101 °C - DIN 51758

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Béatrice Hechler et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 232-243 (2005-03-29)
Our aim was to determine whether the newly described P2X1 antagonist NF449 [4,4',4'',4'''-(carbonylbis(imino-5,1,3-benzenetriylbis(carbonylimino)))tetrakis-benzene-1,3-disulfonic acid octasodium salt] could selectively antagonize the platelet P2X1 receptor and how it affected platelet function. NF449 inhibited alpha,beta-methyleneadenosine 5'-triphosphate-induced shape change (IC50 = 83 +/- 13
Meenakshisundaram Sankar et al.
ACS nano, 6(8), 6600-6613 (2012-07-10)
We report a convenient excess anion modification and post-reduction step to the impregnation method which permits the reproducible preparation of supported bimetallic AuPd nanoparticles having a tight particle size distribution comparable to that found for sol-immobilization materials but without the
Luis E Santos-Figueroa et al.
Chemical Society reviews, 42(8), 3489-3613 (2013-02-13)
This review focuses on examples reported in the years 2010-2011 dealing with the design of chromogenic and fluorogenic chemosensors or reagents for anions.
I H Blank et al.
The Journal of investigative dermatology, 85(6), 522-526 (1985-12-01)
Although it is known that benzene may be absorbed from inhaled air, the amount that may enter the system by percutaneous absorption is less well established. We have measured the penetration of benzene through human abdominal skin in vitro from
Kelly Roberta Francisco et al.
Langmuir : the ACS journal of surfaces and colloids, 28(41), 14531-14539 (2012-09-13)
Polymers formed by the self-assembly of a bis(urea)-based polymer, 2,4-bis(2-ethylhexylureido)toluene (EHUT), in organic solvents such as octane are promising systems with remarkable rheological properties. This is the first self-assembled polymer recently reported as a hydrodynamic drag reducer for hydrocarbons. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service