Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W200298

Sigma-Aldrich

Acetaldehyde solution

40 wt. % in benzyl benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHO
CAS Number:
Molecular Weight:
44.05
FEMA Number:
2003
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.001

availability

available only in EU

concentration

40 wt. % in benzyl benzoate

refractive index

n20/D 1.4540

density

0.868 g/mL at 20 °C
0.956 g/mL at 25 °C

SMILES string

[H]C(C)=O

InChI

1S/C2H4O/c1-2-3/h2H,1H3

InChI key

IKHGUXGNUITLKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service