All Photos(1)
About This Item
Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
Beilstein/REAXYS Number:
956566
MDL number:
availability
available only in EU
concentration
45 wt. % in H2O
SMILES string
CN(C)C
InChI
1S/C3H9N/c1-4(2)3/h1-3H3
InChI key
GETQZCLCWQTVFV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Werner J Geldenhuys et al.
Bioorganic & medicinal chemistry letters, 20(16), 4870-4877 (2010-07-20)
The high affinity neuronal choline transporter (CHT1) is responsible for the uptake of choline into the pre-synaptic terminal of cholinergic neurons. Considering our past experience with modeling the blood-brain barrier choline transporter (BBBCHT) as drug delivery vector to the CNS
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service