T35009
L-Thyroxine sodium salt pentahydrate
99%
Synonym(s):
3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine sodium salt, Sodium levothyroxine, T4
About This Item
assay
99%
optical activity
[α]20/D −6.6°, c = 3 in 1 M NaOH, C2H5OH, 1:2
mp
207-210 (dec.) (lit.)
SMILES string
O.O.O.O.O.[Na+].N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C([O-])=O
InChI
1S/C15H11I4NO4.Na.5H2O/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23;;;;;;/h1-2,4-5,12,21H,3,20H2,(H,22,23);;5*1H2/q;+1;;;;;/p-1/t12-;;;;;;/m0....../s1
InChI key
JMHCCAYJTTWMCX-QWPJCUCISA-M
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
signalword
Danger
hcodes
Hazard Classifications
STOT RE 1
target_organs
Thyroid,Cardio-vascular system,Kidney
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service