Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R406

Sigma-Aldrich

Resorcinol

reagent grade, 98%

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein/REAXYS Number:
906905
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

assay

98%

form

powder or flakes

autoignition temp.

1126 °F

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

solubility

soluble 717 g/L at 25 °C

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1 Oral - STOT SE 2 Oral

target_organs

Central nervous system,Blood, Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

260.6 °F - closed cup

flash_point_c

127 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linfeng Chen et al.
Haematologica, 105(5), 1361-1368 (2019-09-01)
B-cell receptor (BCR) signaling pathway components represent promising treatment targets in multiple B-cell malignancies including diffuse large B-cell lymphoma (DLBCL). In in vitro and in vivo model systems, a subset of DLBCLs depend upon BCR survival signals and respond to
M Sochalska et al.
Cell death and differentiation, 23(4), 628-639 (2015-10-10)
Bcl2 family proteins control mitochondrial apoptosis and its members exert critical cell type and differentiation stage-specific functions, acting as barriers against autoimmunity or transformation. Anti-apoptotic Bcl2a1/Bfl1/A1 is frequently deregulated in different types of blood cancers in humans but its physiological
Nan-Lin Wu et al.
The Journal of investigative dermatology, 136(1), 192-201 (2016-01-15)
Spleen tyrosine kinase (Syk), a nonreceptor tyrosine kinase, was initially identified as a crucial regulator in proximal immunoreceptor signaling. Additional studies have revealed its pleiotropic roles, and drugs targeting Syk are under development for inflammatory diseases. Syk expression in the
Vittorio Abbonante et al.
The Journal of biological chemistry, 288(23), 16738-16746 (2013-03-27)
Growing evidence demonstrates that extracellular matrices regulate many aspects of megakaryocyte (MK) development; however, among the different extracellular matrix receptors, integrin α2β1 and glycoprotein VI are the only collagen receptors studied in platelets and MKs. In this study, we demonstrate
Cecilie F Kjelgaard-Petersen et al.
Biochemical pharmacology, 165, 91-98 (2018-07-31)
Currently, there are no disease-modifying osteoarthritis drugs (DMOADs) approved for osteoarthritis. It is hypothesized that a subtype of OA may be driven by inflammation and may benefit from treatment with anti-inflammatory small molecule inhibitors adopted from treatments of rheumatoid arthritis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service