Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R1102

Sigma-Aldrich

4-Oxo-2-thioxo-3-thiazolidinylacetic acid

95%

Synonym(s):

Rhodanine-3-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO3S2
CAS Number:
Molecular Weight:
191.23
Beilstein/REAXYS Number:
149513
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

mp

145-148 °C (lit.)

SMILES string

OC(=O)CN1C(=O)CSC1=S

InChI

1S/C5H5NO3S2/c7-3-2-11-5(10)6(3)1-4(8)9/h1-2H2,(H,8,9)

InChI key

JGRMXPSUZIYDRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen-Hua Chen et al.
European journal of medicinal chemistry, 45(12), 5739-5743 (2010-10-05)
With an intention to synergize the anti-bacterial activity of chalcones and rhodanine-3-acetic acid, several hybrid compounds possessing chalcone and rhodanine-3-acetic acid moieties were synthesized and tested for their anti-bacterial activity. Some compounds presented great anti-microbial activities against Gram-positive bacteria (including
Ana Martinez et al.
Journal of medicinal chemistry, 48(23), 7103-7112 (2005-11-11)
The 2,4-disubstituted thiadiazolidinones (TDZD) are described as the first ATP-noncompetitive GSK-3 inhibitors. Following an SAR study about TDZD, different structural modifications in the heterocyclic ring aimed to test the influence of each heteroatom on the biological study are here reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service