P2886
Phenoxyacetic acid N-hydroxysuccinimide ester
practical grade
Synonym(s):
Succinimidyl phenoxyacetate
About This Item
Recommended Products
grade
practical grade
impurities
10-20% Free N-hydroxysuccinimide
storage temp.
2-8°C
SMILES string
O=C(COc1ccccc1)ON2C(=O)CCC2=O
InChI
1S/C12H11NO5/c14-10-6-7-11(15)13(10)18-12(16)8-17-9-4-2-1-3-5-9/h1-5H,6-8H2
InChI key
SMBVSRKAVRAVKU-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service