Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P13606

Sigma-Aldrich

2-Phenylethanol

≥99%

Synonym(s):

β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2OH
CAS Number:
Molecular Weight:
122.16
Beilstein/REAXYS Number:
1905732
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.21 (vs air)

vapor pressure

1 mmHg ( 58 °C)

assay

≥99%

refractive index

n20/D 1.5317 (lit.)

bp

219-221 °C/750 mmHg (lit.)

mp

−27 °C (lit.)

density

1.020 g/mL at 20 °C (lit.)

SMILES string

OCCc1ccccc1

InChI

1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

WRMNZCZEMHIOCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaspreet Singh et al.
Biochimica et biophysica acta, 1831(4), 747-758 (2013-01-16)
X-linked adrenoleukodystrophy (X-ALD) is a peroxisomal disorder caused by mutations in the ABCD1 gene. Accumulation of very long chain fatty acids (VLCFA) that have been attributed to reduced peroxisomal VLCFA β-oxidation activity are the hallmark of the disease. Overexpression of
Kartick C Pramanik et al.
Carcinogenesis, 34(9), 2061-2070 (2013-05-04)
Melanoma is highly metastatic and resistant to chemotherapeutic drugs. Our previous studies have demonstrated that caffeic acid phenethyl ester (CAPE) suppresses the growth of melanoma cells and induces reactive oxygen species generation. However, the exact mechanism of the growth suppressive
M M W Etschmann et al.
Applied microbiology and biotechnology, 59(1), 1-8 (2002-06-20)
2-Phenylethanol (2-PE) is an important flavour and fragrance compound with a rose-like odour. Most of the world's annual production of several thousand tons is synthesised by chemical means but, due to increasing demand for natural flavours, alternative production methods are
Li-Zhen Wu et al.
Journal of pharmaceutical and biomedical analysis, 75, 55-63 (2013-01-15)
Ultra high-performance liquid chromatography coupled with electrospray ionization quadrupole time-of-flight tandem mass spectrometry (UHPLC/ESI-Q-TOF MS/MS) was used to investigate the MS fragmentation behaviors of flavone C-glycosides present in the extracts of five Trollius species. In this study, the primary MS
Pradeep Dewapriya et al.
Food chemistry, 141(2), 1147-1157 (2013-06-26)
Experimental evidence suggests that tyrosol [2-(4-hydroxyphenyl)ethanol] exhibits potent protective activities against several pathogeneses. In this study, we evaluated the protective effect of tyrosol against 1-methyl-4-phenylpyridinium (MPP(+))-induced CATH.a neuron cell death. Tyrosol dose-dependently protected CATH.a cells from MPP(+)-induced cell death and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service