Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N7850

Sigma-Aldrich

Nicotinic acid

98%

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein/REAXYS Number:
109591
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

98%

mp

236-239 °C (lit.)

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

379.4 °F - closed cup

flash_point_c

193 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanda C Lakey et al.
Journal of clinical lipidology, 7(2), 178-181 (2013-02-19)
Multiple cholesterol-reducing therapies have been shown to induce the regression of tendon xanthoma in patients with familial hypercholesterolemia. We present 3 cases of adverse reactions in Achilles tendon xanthomas after the addition of niacin and bile acid sequestrants to ongoing
Niacin therapy lives for another day-maybe?
Carl J Lavie et al.
Journal of the American College of Cardiology, 61(21), 2197-2198 (2013-04-02)
Drugs for hypertriglyceridemia.
The Medical letter on drugs and therapeutics, 55(1411), 17-19 (2013-03-08)
Rachel L Edwards et al.
Infection and immunity, 81(3), 945-955 (2013-01-16)
In response to environmental fluctuations or stresses, bacteria can activate transcriptional and phenotypic programs to coordinate an adaptive response. The intracellular pathogen Legionella pneumophila converts from a noninfectious replicative form to an infectious transmissive form when the bacterium encounters alterations
N Nakajima et al.
Clinical toxicology (Philadelphia, Pa.), 51(5), 451-454 (2013-05-07)
Erythromelalgia is a rare disorder characterized by reddening, severe burning pain, and swelling of the extremities. Food poisoning by Clitocybe acromelalga, a poisonous mushroom, is known to induce erythromelalgia; however, its treatment protocol remains unclear. We describe here three cases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service