Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M54600

Sigma-Aldrich

2-(Methylthio)benzimidazole

97%

Synonym(s):

2-(Methylmercapto)benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2S
CAS Number:
Molecular Weight:
164.23
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

202-205 °C (lit.)

SMILES string

CSc1nc2ccccc2[nH]1

InChI

1S/C8H8N2S/c1-11-8-9-6-4-2-3-5-7(6)10-8/h2-5H,1H3,(H,9,10)

InChI key

OCKJFOHZLXIAAT-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodan Chang et al.
American journal of translational research, 8(11), 4694-4704 (2016-12-03)
This study aimed to investigate the potential molecular mechanism underlying the T3 induced vascular calcification and phenotype transformation of vascular smooth muscle cells (VSMCs). Rat thoracic aortic smooth muscle cells (A7r5) were cultured in vitro and randomly assigned into normal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service