Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3701

Sigma-Aldrich

2-Mercaptoethanol

98%

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Beilstein/REAXYS Number:
773648
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

2.69 (vs air)

vapor pressure

1 mmHg ( 20 °C)

assay

98%

expl. lim.

18 %

concentration

14.3 M (pure liquid)

refractive index

n20/D 1.500 (lit.)

bp

157 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BME is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for SDS-PAGE at a concentration of 5%. Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on SDS-PAGE. Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure.
Charge transfer agent for the preparation of polyvinyl chloride.[1] Synthon for the preparation of 1,3-oxathiolanes.[2][3]

Other Notes

This product has been replaced by M6250-ALDRICH | 2-Mercaptoethanol ≥99.0%

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 2 Oral

target_organs

Liver,Heart

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

165.2 °F - closed cup

flash_point_c

74 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Journal of Applied Polymer Science. Applied Polymer Symposium, 51, 555-555 (1994)
Indian J. Chem. B, 32, 1266-1266 (1993)
The Journal of Organic Chemistry, 59, 1053-1053 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service