Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M27387

Sigma-Aldrich

(R)-(+)-3-Methyladipic acid

96%

Synonym(s):

(+)-3-Methylhexanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(CH3)CH2CO2H
CAS Number:
Molecular Weight:
160.17
Beilstein/REAXYS Number:
4661826
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

optical activity

[α]21/D +7.2°, c = 5 in methanol

bp

230 °C/30 mmHg (lit.)

mp

81-84 °C (lit.)

SMILES string

C[C@H](CCC(O)=O)CC(O)=O

InChI

1S/C7H12O4/c1-5(4-7(10)11)2-3-6(8)9/h5H,2-4H2,1H3,(H,8,9)(H,10,11)/t5-/m1/s1

InChI key

SYEOWUNSTUDKGM-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service