M26054
O-Methylserotonin hydrochloride
97%
Synonym(s):
3-(2-Aminoethyl)-5-methoxyindole hydrochloride, 5-Methoxytryptamine hydrochloride, Mexamine hydrochloride
About This Item
assay
97%
mp
245-250 °C (dec.) (lit.)
SMILES string
Cl.COc1ccc2[nH]cc(CCN)c2c1
InChI
1S/C11H14N2O.ClH/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11;/h2-3,6-7,13H,4-5,12H2,1H3;1H
InChI key
TXVAYRSEKRMEIF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service