Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I5567

Sigma-Aldrich

Indole-3-pyruvic acid

98%

Synonym(s):

3-(3-Indolyl)-2-oxopropanoic acid, 3-Indolylpyruvic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO3
CAS Number:
Molecular Weight:
203.19
Beilstein/REAXYS Number:
172966
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

215 °C (dec.) (lit.)

SMILES string

OC(=O)C(=O)Cc1c[nH]c2ccccc12

InChI

1S/C11H9NO3/c13-10(11(14)15)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,12H,5H2,(H,14,15)

InChI key

RSTKLPZEZYGQPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Han et al.
Journal of medicinal chemistry, 52(9), 2786-2793 (2009-04-03)
Human kynurenine aminotransferase I (hKAT I) catalyzes the formation of kynurenic acid, a neuroactive compound. Here, we report three high-resolution crystal structures (1.50-1.55 A) of hKAT I that are in complex with glycerol and each of two inhibitors of hKAT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service