I12654
Iproniazid phosphate salt
97%
Synonym(s):
Isonicotinic acid 2-isopropylhydrazide phosphate salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H13N3O · H3O4P
CAS Number:
Molecular Weight:
277.21
MDL number:
Recommended Products
assay
97%
SMILES string
OP(O)(O)=O.CC(C)NNC(=O)c1ccncc1
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Non-selective MAO-A/B inhibitor.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service