H42001
N-Methyl-3-piperidinol
98%
Synonym(s):
1-Methyl-3-piperidinol, 3-Hydroxy-1-methylpiperidine
About This Item
Recommended Products
assay
98%
refractive index
n20/D 1.474 (lit.)
bp
76-78 °C/11 mmHg (lit.)
density
0.999 g/mL at 25 °C (lit.)
SMILES string
CN1CCCC(O)C1
InChI
1S/C6H13NO/c1-7-4-2-3-6(8)5-7/h6,8H,2-5H2,1H3
InChI key
UKANCZCEGQDKGF-UHFFFAOYSA-N
Application
- Optimization of Novobiocin scaffold to produce antitumor agents
- Formation of carbamates and N-alkylimidazoles
Reactant for synthesis of:
- Pyridine derivatives as CDK5 inhibitors
- Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors
- Amino phosphite ligands
- Mexiletine enantiomers by nucleophilic aromatic substitution
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
158.0 °F - closed cup
flash_point_c
70 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service