Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G6201

Sigma-Aldrich

Glycine

reagent grade, 98%

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein/REAXYS Number:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209

grade

reagent grade

assay

98%

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva S Schaffert et al.
Bioorganic & medicinal chemistry, 19(21), 6492-6504 (2011-09-24)
1,5-Disubstituted and 5-monosubstituted aminomethyltetrazole derivatives derived from glycine were synthesized employing a TMSN(3)-modified variant of the Ugi reaction as a key step. All compounds were evaluated regarding their inhibitory potency and subtype selectivity at the four murine GABA transporter subtypes
Guifeng Kang et al.
Bioorganic & medicinal chemistry letters, 20(20), 6157-6160 (2009-05-24)
Aimed at the chemotherapy of chronic pain two kinds of analgesic pharmacophores, substituted purine and Gly-AA-OBzl, were coupled via a five-step-reaction procedure and 19 novel conjugates N-[2-chloro-9-(tetrahydropyran-2-yl)-9H-purin-6-yl]-N-cyclopropylglycylamino acid benzylesters were provided. On mouse-tail flick model their in vivo analgesic activities
Patrick Maillard et al.
PloS one, 6(10), e26637-e26637 (2011-11-01)
A distinctive feature of HCV is that its life cycle depends on lipoprotein metabolism. Viral morphogenesis and secretion follow the very low-density lipoprotein (VLDL) biogenesis pathway and, consequently, infectious HCV in the serum is associated with triglyceride-rich lipoproteins (TRL). Lipoprotein
Maria Luisa Gelmi et al.
Journal of medicinal chemistry, 50(9), 2245-2248 (2007-04-07)
New 3-O-glycosyl-3-demethylthiocolchicines containing natural and unnatural sugar moieties were prepared and tested on gamma-aminobutyric acid (GABA) and strychnine-sensitive glycine receptors present in rat brain and spinal cord. Two different synthetic approaches were used with the readily available 3-O-demethylthiocolchicine (1b) and
Maria Luisa Gelmi et al.
Journal of medicinal chemistry, 49(18), 5571-5577 (2006-09-01)
A novel class of 3-demethoxy-3-glycosylaminothiocolchicines (7) was prepared and tested for muscle relaxant activity. The syntheses were performed starting from the new 3-amino-3-demethoxythiocolchicine (5) prepared in good yield from 3-O-demethylthiocolchicine (1c) using the Buchwald-Hartwig reaction. The condensation of 5 with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service