Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G4705

Sigma-Aldrich

L-Glutathione reduced

98%

Synonym(s):

γ-L-Glutamyl-L-cysteinyl-glycine, GSH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH(CO2H)CH2CH2CONHCH(CH2SH)CONHCH2CO2H
CAS Number:
Molecular Weight:
307.32
Beilstein/REAXYS Number:
1729812
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

optical activity

[α]25/D −17.0°, c = 2 in H2O

mp

192-195 °C (dec.) (lit.)

SMILES string

N[C@@H](CCC(=O)N[C@@H](CS)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C10H17N3O6S/c11-5(10(18)19)1-2-7(14)13-6(4-20)9(17)12-3-8(15)16/h5-6,20H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)/t5-,6-/m0/s1

InChI key

RWSXRVCMGQZWBV-WDSKDSINSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

γ-Glu-Cys-Gly

Application

Biological antioxidant[1] that has been used with antitumor agents to enhance activity[2] or to reduce side effects.[3]
May be used at 5-10 mM to elute glutathione S-transferase (GST) from glutathione agarose.

Biochem/physiol Actions

Endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst. Glutathione-S-transferase catalyzes the formation of glutathione thioethers with xenobiotics, leukotrienes, and other molecules that have an electrophilic center. Glutathione also forms disulfide bonds with cysteine residues in proteins. Via these mechanisms, it can have the paradoxical effect of reducing the efficacy of anti-cancer agents.

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 116, 6537-6537 (1994)
J H Lee et al.
Cancer research, 54(9), 2398-2403 (1994-05-01)
7-N-((2-([2-(gamma-L-Glutamylamino)ethyl]dithio)ethyl))mitomycin C (KW-2149) is an analogue of mitomycin C (MMC) and has prominent activities against various tumors. We studied the antitumor effects of KW-2149 in MMC-resistant variants of human colon carcinoma HT-29 (HT-29/MMC) and mouse hepatoma Hepa-I (C4, B13NBii1) cells
J. Optim. Nutr., 2, 173-173 (1993)
Mario Faúndez et al.
Antimicrobial agents and chemotherapy, 52(5), 1837-1839 (2008-03-12)
L-buthionine (S,R)-sulfoximine (BSO) at a dose of 220 mg/kg of body weight/day showed an anti-Trypanosoma cruzi effect in infected mice, increasing their survival rate and decreasing the parasitemias and parasite burden in the hearts. Treatment with BSO plus nifurtimox caused
Minakshi C Gurbhele-Tupkar et al.
Bioorganic & medicinal chemistry, 16(5), 2579-2590 (2007-12-11)
Almost all therapeutic proteins and most extracellular proteins contain disulfide bonds. The production of these proteins in bacteria or in vitro is challenging due to the need to form the correctly matched disulfide bonds during folding. One important parameter for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service