G10903
Guaiacol
98%
Synonym(s):
2-Methoxyphenol, Catechol monomethyl ether, Pyrocatechol monomethyl ether
About This Item
Recommended Products
vapor density
4.27 (vs air)
vapor pressure
0.11 mmHg ( 25 °C)
assay
98%
refractive index
n20/D 1.543 (lit.)
bp
205 °C (lit.)
mp
26-29 °C (lit.)
density
1.129 g/mL at 25 °C (lit.)
SMILES string
COc1ccccc1O
InChI
1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3
InChI key
LHGVFZTZFXWLCP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
194.0 °F - closed cup
flash_point_c
90 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service