Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D93902

Supelco

Diethylenetriaminepentaacetic acid

97%

Synonym(s):

(Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(HOOCCH2)2NCH2CH2]2NCH2COOH
CAS Number:
Molecular Weight:
393.35
Beilstein/REAXYS Number:
1810219
EC Number:
MDL number:
UNSPSC Code:
12000000

assay

97%

mp

219-220 °C (lit.)

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O

InChI

1S/C14H23N3O10/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27)

Inchi Key

QPCDCPDFJACHGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

flash_point_f

392.0 °F - closed cup

flash_point_c

200 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolyn Y Ho et al.
JACC. Heart failure, 3(2), 180-188 (2014-12-30)
The study sought to assess the safety, feasibility, and effect of diltiazem as disease-modifying therapy for at-risk hypertrophic cardiomyopathy (HCM) mutation carriers. HCM is caused by sarcomere mutations and characterized by left ventricular hypertrophy (LVH) with increased risk of heart failure
Nikki Rink et al.
Pediatric nephrology (Berlin, Germany), 30(8), 1327-1336 (2015-04-10)
The current glomerular filtration rate (GFR) equation (CKiD) may be less accurate in adolescents and children with higher GFR. This was a retrospective study (n = 161, 247 (99)mTc DTPA-GFRs). Six equations were evaluated for bias, accuracy, and low GFR diagnosis: (1)
Sébastien Leguay et al.
Inorganic chemistry, 51(23), 12638-12649 (2012-11-17)
Complexation of trivalent actinides with DTPA (diethylenetriamine pentaacetic acid) was studied as a function of pcH and temperature in (Na,H)Cl medium of 0.1 M ionic strength. Formation constants of both complexes AnHDTPA(-) and AnDTPA(2-) (where An stands for Am, Cm
Janice S C Chew-Harris et al.
Annals of clinical biochemistry, 50(Pt 1), 39-46 (2012-11-07)
It is well known that plasma creatinine concentration is affected by muscle mass, while some studies have suggested cystatin C is affected by body mass index (BMI). Our aim was to assess the effects of lean versus fat mass on
Vincent Cutillas et al.
Antimicrobial agents and chemotherapy, 59(1), 310-316 (2014-10-29)
Clinical studies have shown that integrase strand transfer inhibitors (INSTIs) can be used effectively against HIV-1 infection. To date, no resistance substitution has been found in INSTI-naive patients treated with the new integrase inhibitor dolutegravir (DTG). In a recent selection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service