Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D61806

Sigma-Aldrich

2,2-Dichloroethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHCH2OH
CAS Number:
Molecular Weight:
114.96
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

146 °C (lit.)

density

1.404 g/mL at 25 °C (lit.)

SMILES string

OCC(Cl)Cl

InChI

1S/C2H4Cl2O/c3-2(4)1-5/h2,5H,1H2

InChI key

IDJOCJAIQSKSOP-UHFFFAOYSA-N

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

172.4 °F - closed cup

flash_point_c

78 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H K Bhat et al.
Toxicology letters, 59(1-3), 203-211 (1991-12-01)
Chloroethanols are toxic chemicals used in industry and also formed as a result of the metabolism of several widely used halogenated hydrocarbons. The effect of 2-chloroethanol (CE), 2,2-dichloroethanol (DCE) and 2,2,2-trichloroethanol (TCE) on rat liver mitochondrial respiration was studied. Rat

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service