Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D47404

Sigma-Aldrich

2,6-Di-tert-butyl-4-methylphenol

99%

Synonym(s):

2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein/REAXYS Number:
1911640
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

assay

99%

autoignition temp.

878 °F

bp

265 °C (lit.)

mp

69-73 °C (lit.)

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Antioxidant

replaced by

Product No.
Description
Pricing

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

260.6 °F - open cup

flash_point_c

127 °C - open cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Vijesh et al.
European journal of medicinal chemistry, 46(11), 5591-5597 (2011-10-05)
In the present study two new series of Hantzsch 1,4-dihydropyridine derivatives (1,4-DHPs) containing substituted pyrazole moiety (4a-f and 5a-f) were synthesized by the reaction of 3-aryl-1H-pyrazole-4-carbaldehydes with 1,3-dicarbonylcompounds (ethylacetoacetate and methylacetoacetate) and ammonium acetate. The newly synthesized compounds were characterized
Gen-ichiro Arimura et al.
Plant physiology, 146(3), 965-973 (2008-01-01)
Continuous mechanical damage initiates the rhythmic emission of volatiles in lima bean (Phaseolus lunatus) leaves; the emission resembles that induced by herbivore damage. The effect of diurnal versus nocturnal damage on the initiation of plant defense responses was investigated using
Metabolism and pulmonary toxicity of butylated hydroxytoluene (BHT).
H Witschi et al.
Pharmacology & therapeutics, 42(1), 89-113 (1989-01-01)
A M Malkinson
Environmental mutagenesis, 5(3), 353-362 (1983-01-01)
Although the average American's daily consumption of BHT can be measured in milligrams, there are numerous reports that BHT causes organ damage in laboratory animals. Only a few genotoxic effects of BHT have been reported, however, including mutagenicity in the
Butylated hydroxytoluene (BHT): a review.
H Babich
Environmental research, 29(1), 1-29 (1982-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service