Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D1653

Sigma-Aldrich

Decanoic acid

96%

Synonym(s):

1-Nonanecarboxylic acid, Acid C10, C10:0, Capric acid, NSC 5025

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOH
CAS Number:
Molecular Weight:
172.26
Beilstein/REAXYS Number:
1754556
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

plant (palm)

vapor pressure

15 mmHg ( 160 °C)

assay

96%

form

solid or liquid

bp

268-270 °C (lit.)

mp

27-32 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCC(O)=O

InChI

1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)

InChI key

GHVNFZFCNZKVNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

296.6 °F - closed cup

flash_point_c

147 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Van Immerseel et al.
Applied and environmental microbiology, 70(6), 3582-3587 (2004-06-09)
The most common source of Salmonella infections in humans is food of poultry origin. Salmonella enterica serovar Enteritidis has a particular affinity for the contamination of the egg supply. In this study, the medium-chain fatty acids (MCFA), caproic, caprylic, and
T M Lima et al.
Toxicology in vitro : an international journal published in association with BIBRA, 16(6), 741-747 (2002-11-09)
The fatty acids have an important role in the control of leukocyte metabolism and function. Higher concentrations of certain fatty acids, particularly polyunsaturated fatty acids (PUFAs) and volatile fatty acids, can cause cell death via apoptosis or, when concentrations are
Hiroyuki Kaiya et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 135(3), 421-429 (2003-07-02)
We have identified ghrelin and cDNA encoding precursor protein from the stomach of a euryhaline tilapia, Oreochromis mossambicus. The sequence of 20-amino acid tilapia ghrelin is GSSFLSPSQKPQNKVKSSRI. The third serine residue was modified by n-decanoic acid. The carboxyl-terminal end of
M Duran et al.
The Journal of pediatrics, 107(3), 397-404 (1985-09-01)
Five patients aged 7 to 21 months are described who developed attacks of coma after a short prodromal illness with diarrhea or vomiting or both. Four had concomitant hypoglycemia, and all had hypoketonemia, with excessive urinary excretion of medium-chain dicarboxylic
Morteza Moradi et al.
Journal of chromatography. A, 1229, 30-37 (2012-02-07)
A new, efficient and environmentally friendly method for the analysis of parabens as model compounds was developed using solidified floating vesicular coacervative drop microextraction (SFVCDME). A supramolecular solvent consisting of vesicles of decanoic acid in the nano- and microscale regimes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service