Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D161004

Sigma-Aldrich

2,5-Dimethyl-2,4-hexadiene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH=C(CH3)2
CAS Number:
Molecular Weight:
110.20
Beilstein/REAXYS Number:
1733342
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.8 (vs air)

vapor pressure

26.9 mmHg ( 37.7 °C)
7.2 mmHg ( 20 °C)

assay

98%

autoignition temp.

559 °F

refractive index

n20/D 1.476 (lit.)

bp

132-134 °C (lit.)

mp

11-14 °C (lit.)

density

0.773 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C/C=C(\C)C

InChI

1S/C8H14/c1-7(2)5-6-8(3)4/h5-6H,1-4H3

InChI key

DZPCYXCBXGQBRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

84.2 °F - closed cup

flash_point_c

29 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service