Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D159409

Sigma-Aldrich

3,3-Dimethylglutaric acid

98%

Synonym(s):

3,3-Dimethylpentanedioic acid, NSC 14987, NSC 49114

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2C(CH3)2CH2COOH
CAS Number:
Molecular Weight:
160.17
Beilstein/REAXYS Number:
1765439
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

mp

100-102 °C (lit.)

SMILES string

CC(C)(CC(O)=O)CC(O)=O

InChI

1S/C7H12O4/c1-7(2,3-5(8)9)4-6(10)11/h3-4H2,1-2H3,(H,8,9)(H,10,11)

InChI key

DUHQIGLHYXLKAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Reactant involved in:
  • Cyclodehydration of diols
  • Synthesis of conjugates of betulin derivatives used as anti-HIV agents
  • Preparation of dimeric peptide antagonists of IgG-FcRn interaction
  • Microwave-assisted protection of glutaraldehyde
  • Synthesis of glycyrrhetinic acid derivatives for proteasome inhibition
  • Catalytic, asymmetric transannular aldolizations

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service