Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D144207

Sigma-Aldrich

1-Dimethylamino-2-propanol

99%

Synonym(s):

N,N-Dimethylisopropanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2N(CH3)2
CAS Number:
Molecular Weight:
103.16
Beilstein/REAXYS Number:
1209244
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.6 (vs air)

vapor pressure

8 mmHg ( 20 °C)

assay

99%

refractive index

n20/D 1.419 (lit.)

bp

121-127 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

SMILES string

CC(O)CN(C)C

InChI

1S/C5H13NO/c1-5(7)4-6(2)3/h5,7H,4H2,1-3H3

InChI key

NCXUNZWLEYGQAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

78.8 °F - DIN 51755 Part 1

flash_point_c

26 °C - DIN 51755 Part 1

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaimie L Gosselin et al.
The journal of physical chemistry. A, 110(12), 4251-4255 (2006-03-24)
The energy flow and fragmentation dynamics of N,N-dimethylisopropylamine (DMIPA) upon excitation to the 3p Rydberg states has been investigated with use of time-resolved photoelectron and mass spectrometry. The 3p states are short-lived, with a lifetime of 701 +/- 45 fs.
The metabolism and excretion in man of NN-dimethylamino-isopropanol and p-acetamido-benzoic acid after administration of isoprinosine.
P Nielsen et al.
The Journal of pharmacy and pharmacology, 33(8), 549-550 (1981-08-01)
Xiang Zhang et al.
Journal of mass spectrometry : JMS, 42(2), 218-224 (2006-12-19)
The methyl migration between the protonated N,N-dimethylisopropylamine and other neutral aliphatic amines in the gas phase has been investigated by a hybrid external chemical ionization source ion trap mass spectrometer. Similar reactions have been found in the aqueous solution by
D G Streeter et al.
Drug metabolism and disposition: the biological fate of chemicals, 12(2), 199-203 (1984-03-01)
The principal excretion products derived from radiolabeled N,N-dimethylaminoisopropanol (Dip) and p-acetamidobenzoic acid (PAcBA) components of inosiplex (Isoprinosine) were identified and quantified in urine following single iv and oral administration of the drug in rhesus monkeys. The major metabolite derived from
B Ballantyne et al.
Veterinary and human toxicology, 38(6), 422-426 (1996-12-01)
The acute handling hazards of several alkylalkanolamines were determined by investigating their potential acute toxicity and primary irritancy. Materials studied were N-methylethanolamine (MEA), N, N, -dimethylethanolamine (DMEA), N, N, -dimethylisopropanolamine (DMIPA), N-methyldiethanolamine (MDEA), and tertbutyldiethanolamine (BDEA). All these alkylalkanolamines were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service