Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C85719

Sigma-Aldrich

o-Cresol

reagent grade, 98%

Synonym(s):

2-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein/REAXYS Number:
506917
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

reagent grade

vapor density

3.72 (vs air)

vapor pressure

0.3 mmHg ( 20 °C)

assay

98%

autoignition temp.

1110 °F

expl. lim.

1.47 %, 148 °F

bp

191 °C (lit.)

mp

29-31 °C (lit.)

SMILES string

Cc1ccccc1O

InChI

1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3

InChI key

QWVGKYWNOKOFNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

177.8 °F - closed cup

flash_point_c

81.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hua-Wei Chen et al.
Chemosphere, 69(2), 184-190 (2007-06-05)
This study investigated the effect of controlled periodic illumination on the photonic efficiency and temporal decomposition behavior of o-cresol by UV/TiO2 process in a slurry reactor using light emitting diodes as the light source. Based on the same UV light
María Eugenia Suarez-Ojeda et al.
Chemosphere, 66(11), 2096-2105 (2006-11-11)
This study examines the feasibility of coupling a Catalytic Wet Air Oxidation (CWAO), with activated carbon (AC) as catalyst, and an aerobic biological treatment to treat a high-strength o-cresol wastewater. Two goals are pursued: (a) To determine the effect of
G Koppen et al.
Journal of applied toxicology : JAT, 27(3), 238-246 (2007-01-18)
The present paper deals with the evaluation of a battery of genotoxicity biomarkers in healthy Flemish adolescents and their relation with common pollutants occurring in their life environment. DNA damage as reflected by the comet assay appeared to be most
Sergio Bordel et al.
Applied microbiology and biotechnology, 74(4), 857-866 (2006-12-01)
The influence of toluene concentration on the specific growth rate, cellular yield, specific CO(2), and metabolite production by Pseudomonas putida F1 (PpF1) was investigated. Both cellular yield and specific CO(2) production remained constant at 1.0 +/- 0.1 g biomass dry
Irama Sanchez et al.
Chemosphere, 68(2), 338-344 (2007-02-16)
This work deals with a new abiotic oxidation process designed as a suitable pre-treatment step within a biological depuration of wastewater containing phenol or its derivatives (o-cresol, 2-chlorophenol and p-nitrophenol) or aniline. The reaction was carried out in a stirred

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service